Anti-MRPL15 Rabbit Polyclonal Antibody
Catalog # 76762-744
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Mitochondrial ribosomal protein L15
- Antigen Symbol:MRPL15
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:29088
- Antigen Synonyms:mitochondrial large ribosomal subunit protein uL15m|HSPC145|MRP-L15|L15mt|mitochondrial|RPML7|MRP-L7|39S ribosomal protein L15
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-296 of human MRPL15 (NP_054894.1).
- Purification:Affinity purification
- Cat. No.:76762-744
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: MRPL15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse