Anti-WBSCR16 Rabbit Polyclonal Antibody
76758-080
: Prosci
- Antibody Type:Primary
- Antigen Name:Williams-Beuren syndrome chromosomal region 16 protein
- Antigen Symbol:WBSCR16
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:81554
- Antigen Synonyms:Williams-Beuren syndrome chromosomal region 16 protein|RCC1-like G exchanging factor-like protein|WBSCR16
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-230 of human WBSCR16 (NP_110425.2).
- Purification:Affinity purification
- Cat. No.:76758-080
- Packaging:Vial
Type: Primary
Antigen: WBSCR16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat