Anti-CLEC4M Rabbit Polyclonal Antibody
Catalog # 76758-134
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:C-type lectin domain family 4, member M
- Antigen Symbol:CLEC4M
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:10332
- Antigen Synonyms:CD209L|DC-SIGN2|HP10347|MGC47866|CD299|CLEC4M|DC-SIGNR|MGC129964|DCSIGNR|L-SIGN|LSIGN
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 280-399 of human CLEC4M (NP_055072.3).
- Purification:Affinity purification
- Cat. No.:76758-134
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: CLEC4M
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat