Anti-PCDH8 Rabbit Polyclonal Antibody
Catalog # 76764-490
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Protocadherin 8
- Antigen Symbol:PCDH8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:5100
- Antigen Synonyms:PCDH8|PAPC|ARCADLIN
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 450-750 of human PCDH8 (NP_002581.2).
- Purification:Affinity purification
- Cat. No.:76764-490
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: PCDH8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat