Anti-HMGCS2 Rabbit Polyclonal Antibody
Catalog # 76751-742
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:3-hydroxy-3-methylglutaryl-CoA synthase 2 (mitochondrial)
- Antigen Symbol:HMGCS2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:3158
- Antigen Synonyms:HMGCS2
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 269-508 of human HMGCS2 (NP_005509.1).
- Purification:Affinity purification
- Cat. No.:76751-742
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: HMGCS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat