Anti-CDA Rabbit Polyclonal Antibody
76750-068
: Prosci
- Antibody Type:Primary
- Antigen Name:Cytidine deaminase
- Antigen Symbol:CDA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:978
- Antigen Synonyms:CDD|Small cytidine deaminase|Cytosine nucleoside deaminase|Cytidine deaminase|CDA|Cytidine aminohydrolase
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-146 of human CDA (NP_001776.1).
- Purification:Affinity purification
- Cat. No.:76750-068
- Packaging:Vial
Type: Primary
Antigen: CDA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse