Anti-SLC4A10 Rabbit Polyclonal Antibody
Catalog # 76758-208
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Solute carrier family 4 member 10
- Antigen Symbol:SLC4A10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:57282
- Antigen Synonyms:NCBE|sodium bicarbonate transporter|NBCn2|sodium bicarbonate cotransporter-like|member 1solute carrier family 4|solute carrier family 4|sodium-driven chloride bicarbonate exchanger|member 10
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 640-725 of human SLC4A10 (NP_001171486.1).
- Purification:Affinity purification
- Cat. No.:76758-208
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: SLC4A10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse