Anti-MAN1B1 Rabbit Polyclonal Antibody
Catalog # 76759-192
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Mannosidase alpha class 1B member 1
- Antigen Symbol:MAN1B1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:11253
- Antigen Synonyms:ERManI|MRT15|endoplasmic Reticulum Class I alpha-mannosidase|ER alpha 1,2-mannosidase|MANA-ER|Man9GlcNAc2-specific processing alpha-mannosidase|ERMAN1|endoplasmic reticulum mannosyl-oligosaccharide 1,2-alpha-mannosidase|endoplasmic reticulum mannosyl-oligosaccharide 1,2-alpha-mannosidase 1
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-290 of human MAN1B1 (NP_057303.2).
- Purification:Affinity purification
- Cat. No.:76759-192
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: MAN1B1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat