Anti-EDC3 Rabbit Polyclonal Antibody
Catalog # 76753-962
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Enhancer of mRNA-decapping protein 3
- Antigen Symbol:EDC3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:80153
- Antigen Synonyms:hYjeF_N2|Enhancer of mRNA-decapping protein 3|YJDC|YjeF N-terminal domain-containing protein 2|LSM16|LSM16 homolog|EDC3|YJEFN2|YjeF_N2|YjeF domain-containing protein 1
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 159-508 of human EDC3 (NP_079359.2).
- Purification:Affinity purification
- Cat. No.:76753-962
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: EDC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat