Anti-MAOA Rabbit Polyclonal Antibody
Catalog # 76759-094
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Monoamine oxidase A
- Antigen Symbol:MAOA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:4128
- Antigen Synonyms:MAO-A|MAOA
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human MAOA (NP_000231.1).
- Purification:Affinity purification
- Cat. No.:76759-094
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: MAOA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat