Anti-TNFRSF11A Rabbit Polyclonal Antibody
Catalog # 76763-982
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Tumor necrosis factor receptor superfamily, member 11a, NFKB activator
- Antigen Symbol:TNFRSF11A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:8792
- Antigen Synonyms:TNFRSF11A|ODFR|TRANCE-R|CD265
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 466-616 of human TNFRSF11A (NP_003830.1).
- Purification:Affinity purification
- Cat. No.:76763-982
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: TNFRSF11A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat