Anti-GAG Mouse Monoclonal Antibody [clone: 8G9 ] (Biotinylated)
77124-024
: Prosci
- Antibody Type:Primary
- Antigen Name:gag Pr55(Gag)
- Antigen Symbol:GAG
- Clonality:Monoclonal
- Clone:8G9
- Conjugation:Biotin
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Virus
- Western Blot:Yes
- Size:0.02 mg
- Form:Liquid
- Gene ID:155030
- Antigen Synonyms:HIV-1 p24 Antibody [8G9] :
- Modification:None
- Storage Buffer:HIV-1 p24 Monoclonal antibody is supplied in PBS containing 1% BSA and 0.02% sodium azide.
- Storage Temperature:HIV-1 p24 monoclonal antibody can be stored at 4 °C for three months and −20 °C, stable for up to one year.
- Concentration:1 mg/mL
- Shipping Temperature:20
- Immunogen:Mouse monoclonal HIV-1 p24 antibody was raised against a recombinant full-length HIV-1 p24 protein.
- Purification:HIV-1 p24 Monoclonal antibody is rotein A purified.
- Cat. No.:77124-024
- Packaging:Vial
Type: Primary
Antigen: GAG
Clonality: Monoclonal
Clone: 8G9
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Virus