Anti-Il5 Rat Monoclonal Antibody [clone: [TRFK5]]
Catalog # 77121-700
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:interleukin 5
- Antigen Symbol:Il5
- Clonality:Monoclonal
- Clone:[TRFK5]
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Rat
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Mouse
- Size:0.5 mg
- Form:Liquid
- Gene ID:16191
- Antigen Synonyms:TRF-1|Eo-CSF|IgA-EF|Interleukin-5|BCGF-2|EDF|eosinophil differentiation factor|eosinophil colony stimulating factor|IgA enhancing factor|T-cell replacing factor-1|B cell growth factor-2
- Modification:None
- Storage Buffer:Supplied in borate buffered saline, pH 8.2; No preservatives or amine-containing buffer salts added.
- Storage Temperature:Store vial at 2...8 °C.
- Concentration:0.5 mg/ml
- Shipping Temperature:20
- Immunogen:Semi-purified T cell clone supernatant.
- Cat. No.:77121-700
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: Il5
Clonality: Monoclonal
Clone: [TRFK5]
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse