Anti-PTPRC Mouse Monoclonal Antibody [clone: [F10-89-4]] (PE (Phycoerythrin)-Cy7®)
Catalog # 77125-898
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Receptor-type tyrosine-protein phosphatase C
- Antigen Symbol:PTPRC
- Clonality:Monoclonal
- Clone:[F10-89-4]
- Conjugation:PE (Phycoerythrin)-Cy7®
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Horse,Human
- Size:100 tests
- Form:Liquid
- Gene ID:5788
- Antigen Synonyms:Ly5|T200|LCA
- Modification:None
- Storage Buffer:Supplied in PBS/NaN3 and a stabilizing agent.
- Storage Temperature:Store vial at 2...8 °C.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Purified T cells from human lymph nodes.
- Cat. No.:77125-898
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: [F10-89-4]
Conjugation: PE (Phycoerythrin)-Cy7®
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Horse