Anti-Mouse Monoclonal Antibody [clone: [TCR-2]] (FITC (Fluorescein Isothiocyanate))
77126-552
: Prosci
- Antibody Type:Primary
- Clonality:Monoclonal
- Clone:[TCR-2]
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Turkey,Guinea fowl,Chicken,Peacock
- Size:0.5 mg
- Form:Liquid
- Antigen Synonyms:T3/TCR complex|TCR alpha/beta
- Modification:None
- Storage Buffer:Supplied in PBS/NaN3.
- Storage Temperature:Store vial at 2...8 °C.
- Concentration:0.5 mg/ml
- Shipping Temperature:20
- Immunogen:White Leghorn chick thymocytes and blood mononuclear cells.
- Tested Applications:Complement Mediated Cell Depletion
- Cat. No.:77126-552
- Packaging:Vial
Type: Primary
Antigen:
Clonality: Monoclonal
Clone: [TCR-2]
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chicken, Turkey, Peacock, Guinea Fowl