Anti-FN1 Mouse Monoclonal Antibody [clone: [5G7]] (FITC (Fluorescein Isothiocyanate))
Catalog # 76749-284
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:fibronectin 1
- Antigen Symbol:FN1
- Clonality:Monoclonal
- Clone:[5G7]
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human,Rat
- Western Blot:Yes
- Size:0.1 mg
- Form:Liquid
- Gene ID:2335
- Antigen Synonyms:FN
- Modification:None
- Storage Buffer:Supplied in PBS/NaN3.
- Storage Temperature:Store vial at 2...8 °C.
- Concentration:0.1 mg/ml
- Shipping Temperature:20
- Immunogen:Human plasma fibronectin.
- Cat. No.:76749-284
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: FN1
Clonality: Monoclonal
Clone: [5G7]
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat