Anti-CADM1 Goat Polyclonal Antibody
77120-496
: Prosci
- Antibody Type:Primary
- Antigen Name:cell adhesion molecule 1
- Antigen Symbol:CADM1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human
- Western Blot:Yes
- Size:0.1 mg
- Form:Liquid
- Gene ID:23705
- Antigen Synonyms:tumor suppressor in lung cancer 1|nectin-like protein 2|synCAM1|IGSF4|cell adhesion molecule 1|sgIGSF|member 4|immunoglobulin superfamily|TSLC1/Nectin-like 2/IGSF4|SYNCAM|CADM1|BL2|IGSF4A|DKFZp686F1789|TSLC1|NECL2|ST17|nectin-like 2|Necl-2|sTSLC-1|MGC149785|MGC51880|RA175
- Modification:None
- Storage Buffer:Supplied at 0.5 mg/ml in Tris saline, 0.02% sodium azide, pH7.3 with 0.5% bovine serum albumin.
Aliquot and store at -20°C. Minimize freezing and thawing. - Storage Temperature:Aliquot and store at −20 °C. Minimize freezing and thawing.
- Concentration:500 µg/ml
- Shipping Temperature:20
- Immunogen:The immunogen for this antibody is: C-NLQTQRYLEVQ.
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. No.:77120-496
- Packaging:Vial
Type: Primary
Antigen: CADM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human