Anti-Transferrin receptor 1 Goat Polyclonal Antibody
Catalog # 77120-772
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:transferrin receptor
- Antigen Symbol:Transferrin receptor 1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human
- Western Blot:Yes
- Size:0.1 mg
- Form:Liquid
- Gene ID:7037
- Antigen Synonyms:TRFR|CD71)|T9|TFR1|OTTHUMP00000208525|OTTHUMP00000208523|OTTHUMP00000208524|transferrin receptor protein 1|CD71|transferrin receptor (p90|p90|TFRC|TFR|TR
- Modification:None
- Storage Buffer:Supplied at 0.5 mg/ml in Tris saline, 0.02% sodium azide, pH7.3 with 0.5% bovine serum albumin.
Aliquot and store at -20°C. Minimize freezing and thawing. - Storage Temperature:Aliquot and store at −20 °C. Minimize freezing and thawing.
- Concentration:500 µg/ml
- Shipping Temperature:20
- Immunogen:The immunogen for this antibody is: TRFSLARQVDGDNSH-C.
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. No.:77120-772
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: Transferrin receptor 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human