Anti-SMARCD1 Rabbit Polyclonal Antibody
Catalog # 76762-704
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily D member 1
- Antigen Symbol:SMARCD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:83797
- Antigen Synonyms:BRG1-associated factor 60A|Smarcd1|D15Kz1|SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily D member 1|Baf60a|BAF60A|Protein D15KZ1|60 kDa BRG-1/Brm-associated factor subunit A|SWI/SNF complex 60 kDa subunit
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH7.3.
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-120 of mouse BAF60a (NP_114030.2).
- Purification:Affinity purification.
- Cat. No.:76762-704
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: SMARCD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat