Anti-HOXA2 Rabbit Polyclonal Antibody
76760-280
: Prosci
- Antibody Type:Primary
- Antigen Name:homeobox A2
- Antigen Symbol:HOXA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:3199
- Antigen Synonyms:HOX1K|MCOHI|HOXA2
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH7.3.
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 207-376 of human HOXA2 (NP_006726.1).
- Purification:Affinity purification.
- Cat. No.:76760-280
- Packaging:Vial
Type: Primary
Antigen: HOXA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse