Anti-HACE1 Rabbit Polyclonal Antibody
Catalog # 76761-712
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:HECT domain and ankyrin repeat containing E3 ubiquitin protein ligase 1
- Antigen Symbol:HACE1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:57531
- Antigen Synonyms:HACE1|KIAA1320
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH7.3.
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:A synthetic peptide corresponding to a sequence within amino acids 1-100 of human HACE1 (NP_065822.2).
- Purification:Affinity purification.
- Cat. No.:76761-712
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: HACE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat