Anti-AQP9 Rabbit Polyclonal Antibody
Catalog # 76763-848
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:aquaporin 9
- Antigen Symbol:AQP9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:366
- Antigen Synonyms:HsT17287|SSC1|aquaglyceroporin-9|T17287|AQP-9|small solute channel 1|aquaporin-9
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH7.3.
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:A synthetic peptide corresponding to a sequence within amino acids 50-150 of human AQP9 (NP_066190.2).
- Purification:Affinity purification.
- Cat. No.:76763-848
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: AQP9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat