Anti-SFI1 Rabbit Polyclonal Antibody
76759-262
: Prosci
- Antibody Type:Primary
- Antigen Name:SFI1 centrin binding protein
- Antigen Symbol:SFI1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:9814
- Antigen Synonyms:PPP1R139|protein SFI1 homolog|Sfi1 homolog|regulatory subunit 139|protein phosphatase 1|PISD|hSfi1p|homolog of yeast Sfi1|spindle assembly associated
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH7.3.
- Storage Temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 972-1211 of human SFI1 (NP_055590.2).
- Purification:Affinity purification.
- Cat. No.:76759-262
- Packaging:Vial
Type: Primary
Antigen: SFI1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human