Lifeasible Bacillus thuringiensis Recombinant Cry1Ab from (E. coli)
Catalog # 76683-086
Supplier: Creative Biomart
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein Function:Insecticidal protein
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Bacillus thuringiensis
- Size:1 mg
- Storage Conditions:–80 °C
- Animal-Free:Y
- Protein/Peptide Name:Cry1Ab
- Purity:80%
- Molecular Weight:60 - 70 kDa
- Sequence:MDNNPNINECIPYNCLSNPEVEVLGGERIETGYTPIDISLSLTQFLLSEFVPGAGFVLGLVDIIWGIFGPSQWDAFLVQIEQLINQRIEEFARNQAISRLEGLSNLYQIYAESFREWEADPTNPALREEMRIQFNDMNSALTTAIPLFAVQNYQVPLLSVYVQAANLHLSVLRDVSVFGQRWGFDAATINSRYNDLTRLIGNYTDHAVRWYNTGLERVWGPDSRDWIRYNQFRRELTLTVLDIVSLFPNYDSRTYPIRTVSQLTREIYTNPVLENFDGSFRGSAQGIEGSIRSPHLMDILNSITIYTDAHRGEYYWSGHQIMASPVGFSGPEFTFPLYGTMGNAAPQQRIVAQLGQGVYRTLSSTLYRRPFNIGINNQQLSVLDGTEFAYGTSSNLPSAVYRKSGTVDSLDEIPPQNNNVPPRQGFSHRLSHVSMFRSGFSNSSVSIIRAPMFSWIHRSAEFNNIIPSSQITQIPLTKSTNLGSGTSVVKGPGFTGGDILRRTSPGQISTLRVNITAPLSQRYRVRIRYASTTNLQFHTSIDGRPINQGNFSATMSSGSNLQSGSFRTVGFTTPFNFSNGSSVFTLSAHVFNSGNEVYIDRIEFVPAEVTFEAEYDLERAQKAVNELFTSSNQIGLKTDVTDYHIDQVSNLVECLSDEFCLDEKKELSEKVKHAKRLSDERNLLQDPNFRGINRQLDRGWRGSTDITIQGGDDVFKENYVTLLGTFDECYPTYLYQKIDESKLKAYTRYQLRGYIEDSQDLEIYLIRYNAKHETVNVPGTGSLWPLSAPSPIGKCAHHSHHFSLDIDVGCTD
- Concentration:1 mg/ml
- Shipping Temperature:–80 °C
- Cat. No.:76683-086
Specifications
About this item
Promotes colloidosmotic lysis by binding to the midgut epithelial cells of lepidopteran (Manduca sexta) larvae.
Delivery: Dry ice.