Caspase 6 alpha Peptide
Catalog # 77124-814
Supplier: Prosci
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Synthetic
- Source:
- Species:Human
- Size:0.1 mg
- Environmentally Preferable:
- Storage Conditions:Store at −20 °C. When reconstituted minimize freezing and thawing.
- Gene ID:839
- Protein Synonyms:apoptosis-related cysteine peptidase|caspase 6|apoptotic protease MCH-2|CASP6|caspase-6|MCH2|apoptosis-related cysteine protease
- Protein/Peptide Name:Caspase 6
- Formulation:Lyophilized. Caspase 6 alpH a peptide is supplied as 100 µg of dried peptide. When the peptide is reconstituted in 200 µl water, the concentration will be 0.5 mg/ml.
- Shipping Temperature:RT
- Tested Applications:Blocking
- Cat. No.:77124-814