TNNT3 Peptide
Catalog # 77123-630
Supplier: Prosci
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Synthetic
- Species:HumanRatMouse
- Size:0.1 mg
- Storage Conditions:Store at −20 °C. When reconstituted minimize freezing and thawing.
- Gene ID:7140
- Protein Synonyms:troponin T|beta-TnTF|fast)|troponin T type 3 (skeletal|fast|TNTF|skeletal|troponin-T3|TNNT3|fast skeletal muscle troponin T|fast skeletal muscle|fTnT|beta TnTF
- Protein/Peptide Name:TNNT3
- Formulation:Lyophilized. TNNT3 peptide is supplied as 100 µg of dried peptide. When the peptide is reconstituted in 200 µl water, the concentration will be 0.5 mg/ml.
- Shipping Temperature:RT
- Tested Applications:Blocking
- Cat. No.:77123-630