biotinidase Peptide
Catalog # 77127-172
Supplier: Prosci
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Synthetic
- Species:Human
- Size:0.1 mg
- Storage Conditions:Store at −20 °C. When reconstituted minimize freezing and thawing.
- Gene ID:686
- Protein Synonyms:biotinase|OTTHUMP00000209138|OTTHUMP00000209135|biotinidase|OTTHUMP00000209134|OTTHUMP00000160300|BTD
- Protein/Peptide Name:Biotinidase
- Formulation:Lyophilized. biotinidase peptide is supplied as 100 µg of dried peptide. When the peptide is reconstituted in 200 µl water, the concentration will be 0.5 mg/ml.
- Shipping Temperature:RT
- Tested Applications:Blocking
- Cat. No.:77127-172