AOC3/VAP1 Peptide
Catalog # 77126-406
Supplier: Prosci
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Synthetic
- Species:Human
- Size:0.1 mg
- Storage Conditions:Store at −20 °C. When reconstituted minimize freezing and thawing.
- Gene ID:8639
- Protein Synonyms:copper amine oxidase|copper containing 3|vascular adhesion protein 1|VAP-1|semicarbazide-sensitive amine oxidase|HPAO|copper containing 3 (vascular adhesion protein 1)|SSAO|AOC3|VAP1|amine oxidase
- Protein/Peptide Name:AOC3/VAP1
- Formulation:Lyophilized. AOC3 / VAP1 peptide is supplied as 100 µg of dried peptide. When the peptide is reconstituted in 200 µl water, the concentration will be 0.5 mg/ml.
- Shipping Temperature:RT
- Tested Applications:Blocking
- Cat. No.:77126-406