CLU Peptide
Catalog # 77135-094
Supplier: Prosci
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Synthetic
- Species:Human
- Size:0.1 mg
- Storage Conditions:Store at −20 °C. When reconstituted minimize freezing and thawing.
- Gene ID:1191
- Protein Synonyms:SGP-2|NA1/NA2|KUB1|SP-40|TRPM-2|SGP2|Clusterin|CLU1|APOJ|Apo-J|CLI|APO-J|TRPM2|Aging-associated gene 4 protein|AAG4|CLU2
- Protein/Peptide Name:CLU
- Formulation:Lyophilized. CLU Peptide is supplied as 100 µg of dried peptide. When the peptide is reconstituted in 200 µl water, the concentration will be 0.5 mg/ml.
- Shipping Temperature:RT
- Tested Applications:Blocking
- Cat. No.:77135-094