ARPC1A Peptide
Catalog # 77125-930
Supplier: Prosci
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Synthetic
- Species:Human
- Size:0.1 mg
- Storage Conditions:Store at −20 °C. When reconstituted minimize freezing and thawing.
- Gene ID:10552
- Protein Synonyms:Arc40|SOP2Hs|HEL-68|SOP2-like protein|SOP2L
- Protein/Peptide Name:ARPC1A
- Formulation:Lyophilized. ARPC1A peptide is supplied as 100 µg of dried peptide. When the peptide is reconstituted in 200 µl water, the concentration will be 0.5 mg/ml.
- Shipping Temperature:RT
- Tested Applications:Blocking
- Cat. No.:77125-930