Order Entry
United States
Orders LinkContactUsLinkComponent
Muc5ac Peptide

Muc5ac Peptide

Vial, 0.1 mg
Catalog # 77127-158
Supplier:   Prosci
Supplier #:  42-329P
Muc5ac Peptide
Catalog # 77127-158
Supplier:  Prosci
CAS Number:  
Muc5ac Peptide

Vial, 0.1 mg

CAS Number:  
Price
$106.68
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Vial
  • Size:
    0.1 mg
  • Storage Conditions:
    Store at −20 °C. When reconstituted minimize freezing and thawing.
  • Cat. No.:
    77127-158
  • Shipping Temperature:
    RT
  • Protein/Peptide Type:
    Synthetic
  • Gene ID:
    17833
  • Formulation:
    Lyophilized. Muc5ac peptide is supplied as 100 µg of dried peptide. When the peptide is reconstituted in 200 µl water, the concentration will be 0.5 mg/ml.
  • Tested Applications:
    Blocking
  • Protein Synonyms:
    MGM|2210005L13Rik
  • Protein/Peptide Name:
    Muc5ac
  • Conjugation:
    Unconjugated

Specifications