Human Recombinant B7-H3 (4Ig)/B7-H3b Protein (from HEK293 cells)
Catalog # 77133-280
Supplier: Prosci
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:0.1 mg
- Storage Conditions:Lyophilized protein should be stored at −20 °C or lower for long term storage. Upon reconstitution, working aliquots should be stored at −20 or −70 °C. Avoid repeated freeze-thaw cycles.
- Gene ID:80381
- Protein Synonyms:B7 homolog 3|B7-H3|UNQ309/PRO352|4Ig-B7-H3|PSEC0249|CD276
- Protein/Peptide Name:CD276
- Purity:>95% as determined by SDS-PAGE
- Molecular Weight:48.5 kDa
- Sequence:Gly 27 - Thr 461
- Formulation:Lyophilized. PBS, pH 7.4.
- Shipping Temperature:RT
- Tested Applications:ELISA,WB
- Cat. No.:77133-280