Human Recombinant IL-8/CXCL8 Protein (from HEK293 cells)
Catalog # 77120-644
Supplier: Prosci
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:0.1 mg
- Storage Conditions:Lyophilized protein should be stored at −20 °C or lower for long term storage. Upon reconstitution, working aliquots should be stored at −20 or −70 °C. Avoid repeated freeze-thaw cycles.
- Gene ID:3576
- Protein Synonyms:MDNCF|LYNAP|IL8|NAP-1|GCP1|MONAP|LECT|NAF|CXCL8|LUCT
- Protein/Peptide Name:CXCL8
- Purity:>90% as determined by SDS-PAGE
- Molecular Weight:10.3 kDa
- Sequence:Ser 28 - Ser 99
- Formulation:Lyophilized. PBS, pH 7.4.
- Shipping Temperature:RT
- Tested Applications:ELISA,WB
- Cat. No.:77120-644