Mouse Recombinant Glypican 2/GPC2 Protein (from HEK293 cells)
77131-172
: Prosci
:
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Size:0.1 mg
- Storage Conditions:Lyophilized protein should be stored at −20 °C or lower for long term storage. Upon reconstitution, working aliquots should be stored at −20 or −70 °C. Avoid repeated freeze-thaw cycles.
- Gene ID:71951
- Protein Synonyms:GPC2|Glypican 2
- Protein/Peptide Name:Gpc2
- Purity:>90% as determined by SDS-PAGE
- Molecular Weight:60.6 kDa
- Sequence:His 22 - Ser 556
- Formulation:Lyophilized. PBS, pH 7.4.
- Shipping Temperature:RT
- Tested Applications:ELISA,WB
- Cat. No.:77131-172