Human Recombinant Vitronectin/VTN Protein (from E. coli)
77122-746
: Prosci
:
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:0.1 mg
- Storage Conditions:Lyophilized protein should be stored at −20 °C or lower for long term storage. Upon reconstitution, working aliquots should be stored at −20 or −70 °C. Avoid repeated freeze-thaw cycles.
- Gene ID:7448
- Protein Synonyms:Serum-spreading factor|Vitronectin|VTN|S-protein|V75|VN
- Protein/Peptide Name:VTN
- Purity:>90% as determined by SDS-PAGE
- Molecular Weight:43.2 kDa
- Sequence:Asp 20 -Arg 398
- Formulation:Lyophilized. 20 mM Tris-HCl, Arginine, pH 8.0 .
- Shipping Temperature:RT
- Tested Applications:ELISA,WB
- Cat. No.:77122-746