Anti-TIGIT Recombinant Antibody [Clone: 1B4]
Catalog # 76629-408
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Clonality:Recombinant
- Clone:1B4
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Isotype:IgG lambda
- Reactivity:Mouse
- Size:0.2 mg
- Cross Adsorption:No
- Form:Liquid
- Antigen Synonyms:T cell immunoreceptor with Ig and ITIM domains|WUCAM|VSTM3|V-set and transmembrane domain-containing protein 3|T cell Ig and ITIM domain
- Modification:Unmodified
- Storage Buffer:PBS with 0.02% Proclin 300
- Storage Temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping Temperature:Shipped on blue ice
- Immunogen:This antibody was raised by immunising TIGIT-/- mice with recombinant murine TIGIT tetramers.
- Tested Applications:Blocking
- Cat. No.:76629-408
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: TIGIT
Clonality: Recombinant
Clone: 1B4
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG lambda
Reactivity: Mouse