Anti-Ifngr1 Recombinant Antibody [Clone: GR20]
76630-380
: Prosci
- Antibody Type:Primary
- Antigen Name:interferon gamma receptor 1
- Antigen Symbol:Ifngr1
- Clonality:Recombinant
- Clone:GR20
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Size:0.2 mg
- Cross Adsorption:No
- Neutralisation:Yes
- Form:Liquid
- Gene ID:15979
- Antigen Synonyms:Interferon gamma receptor alpha-chain|Interferon gamma receptor 1|IFN-gamma-R-alpha|CD119|IFN gamma receptor 1|IFN-gamma-R1
- Modification:Unmodified
- Storage Buffer:PBS with 0.02% Proclin 300
- Storage Temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping Temperature:Shipped on blue ice
- Immunogen:This antibody was raised by immunising rats with interferon gamma receptor.
- Tested Applications:Blocking, assay
- Cat. No.:76630-380
- Packaging:Vial
Type: Primary
Antigen: interferon gamma receptor 1
Clonality: Recombinant
Clone: GR20
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG2a kappa
Reactivity: Mouse