Anti-Cmklr1 Recombinant Antibody [Clone: BZ194]
Catalog # 76630-670
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:chemokine-like receptor 1
- Antigen Symbol:Cmklr1
- Clonality:Recombinant
- Clone:BZ194
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Western Blot:Yes
- Size:0.2 mg
- Cross Adsorption:No
- Form:Liquid
- Gene ID:14747
- Antigen Synonyms:DEZ|Dez|Gpcr27|Chemerin receptor|G-protein coupled receptor DEZ|ChemR23|Resolvin E1 receptor|MGC126105|RVER1|CHEMERINR|orphan G-protein coupled receptor|chemokine receptor-like 1|G-protein coupled receptor ChemR23|MGC126106|Chemokine-like receptor 1|CMKLR1
- Modification:Unmodified
- Storage Buffer:PBS with 0.02% Proclin 300
- Storage Temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping Temperature:Shipped on blue ice
- Immunogen:This antibody was raised by immunising Wistar Furth rats with the mCMKLR1 peptide/KLH conjugate. The immunizing amino-terminal mCMKLR1 peptide with the sequence NH2-DSGIYDDEYSDGFGYFVDLEEASPWC-COOH, corresponding to residues residues 8–32 of mCMKLR1, with a nonnative carboxyl-terminal cysteine to facilitate conjugation to keyhole limpet hemocyanin (KLH).
- Tested Applications:Functional assays
- Cat. No.:76630-670
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: chemokine-like receptor 1
Clonality: Recombinant
Clone: BZ194
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG2a kappa
Reactivity: Mouse