Anti-Pdcd1lg2 Recombinant Antibody [Clone: TY25]
Catalog # 76629-458
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:programmed cell death 1 ligand 2
- Antigen Symbol:Pdcd1lg2
- Clonality:Recombinant
- Clone:TY25
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG kappa
- Reactivity:Mouse
- Western Blot:Yes
- Size:0.2 mg
- Cross Adsorption:No
- Form:Liquid
- Gene ID:58205
- Antigen Synonyms:CD273|Pdcd1lg2|B7DC|PDL2|programmed death ligand 2|B7-DC
- Modification:Unmodified
- Storage Buffer:PBS with 0.02% Proclin 300
- Storage Temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping Temperature:Shipped on blue ice
- Immunogen:This antibody was raised by immunising SD rats with B7-DC-transfected RAW264.7 cells and selected on B7-DC-transfected NRK.
- Tested Applications:Blocking
- Cat. No.:76629-458
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: programmed cell death 1 ligand 2
Clonality: Recombinant
Clone: TY25
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG kappa
Reactivity: Mouse