Anti-RTN4RL1 Recombinant Antibody [Clone: M5]
76630-534
: Prosci
- Antibody Type:Primary
- Antigen Name:reticulon 4 receptor like 1
- Antigen Symbol:RTN4RL1
- Clonality:Recombinant
- Clone:M5
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG kappa
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:0.2 mg
- Cross Adsorption:No
- Form:Liquid
- Gene ID:146760
- Antigen Synonyms:Nogo-66 Receptor
- Modification:Unmodified
- Storage Buffer:PBS with 0.02% Proclin 300
- Storage Temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping Temperature:Shipped on blue ice
- Immunogen:(His)6-tagged fragment of rat NgR1 (amino acids 278-473).
- Cat. No.:76630-534
- Packaging:Vial
Type: Primary
Antigen: reticulon 4 receptor like 1
Clonality: Recombinant
Clone: M5
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG kappa
Reactivity: Mouse;Rat;Human