Anti-SEPTIN5 Recombinant Antibody [Clone: SP18]
Catalog # 76629-520
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:septin 5
- Antigen Symbol:SEPTIN5
- Clonality:Recombinant
- Clone:SP18
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Rabbit,Human,Rat
- Western Blot:Yes
- Size:0.2 mg
- Cross Adsorption:No
- Form:Liquid
- Gene ID:5413
- Antigen Synonyms:PNUTL1|CDCREL1|Septin-5|SEPT5|H5|CDCrel-1|CDCREL|CDCREL-1
- Modification:Unmodified
- Storage Buffer:PBS with 0.02% Proclin 300
- Storage Temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Immunoprecipitate from human brain.
- Cat. No.:76629-520
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: septin 5
Clonality: Recombinant
Clone: SP18
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1 kappa
Reactivity: Human;Rat;Rabbit