Anti-STX Recombinant Antibody [Clone: SP6]
Catalog # 76630-148
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Clonality:Recombinant
- Clone:SP6
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Hamster,Pig,Rabbit,Human,Bovine,Guinea pig,Dog,Rat
- Western Blot:Yes
- Size:0.2 mg
- Cross Adsorption:No
- Form:Liquid
- Antigen Synonyms:STX
- Modification:Unmodified
- Storage Buffer:PBS with 0.02% Proclin 300
- Storage Temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synaptic vesicle-containing fractions immunoprecipitated from human brain homogenates.
- Cat. No.:76630-148
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: STX
Clonality: Recombinant
Clone: SP6
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1 kappa
Reactivity: Human;Rat;Hamster;Rabbit;Pig;Dog;Guinea pig;Bovine