Anti-CEA Recombinant Antibody [Clone: Arcitumomab]
76629-272
: Prosci
- Antibody Type:Primary
- Clonality:Recombinant
- Clone:Arcitumomab
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:0.2 mg
- Cross Adsorption:No
- Form:Liquid
- Antigen Synonyms:CEA
- Modification:Unmodified
- Storage Buffer:PBS with 0.02% Proclin 300
- Storage Temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Carcinoembryonic antigen (CEA) describes a set of highly related glycoproteins involved in cell adhesion normally produced in gastrointestinal tissue during fetal development, but only at very low levels in adults.
- Tested Applications:Imaging
- Cat. No.:76629-272
- Packaging:Vial
Type: Primary
Antigen: CEA
Clonality: Recombinant
Clone: Arcitumomab
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1 kappa
Reactivity: Human