Anti-AOC3 Recombinant Antibody [Clone: 2D10]
76629-584
: Prosci
- Antibody Type:Primary
- Antigen Name:amine oxidase copper containing 3
- Antigen Symbol:AOC3
- Clonality:Recombinant
- Clone:2D10
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:0.2 mg
- Cross Adsorption:No
- Form:Liquid
- Gene ID:8639
- Antigen Synonyms:Amine oxidase|copper containing 3|Vascular adhesion protein 1|Membrane primary amine oxidase|Semicarbazide-sensitive|HPAO|copper-containing amine:oxyg|SSAO|AOC3|VAP1|Copper amine oxidase|amine oxidase
- Modification:Unmodified
- Storage Buffer:PBS with 0.02% Proclin 300
- Storage Temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Affinity purified VAP-1 from tonsil stroma of human origin.
- Cat. No.:76629-584
- Packaging:Vial
Type: Primary
Antigen: amine oxidase copper containing 3
Clonality: Recombinant
Clone: 2D10
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1 kappa
Reactivity: Human