Anti-CD52 Recombinant Antibody [Clone: Campath-1H]
Catalog # 76629-250
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:CD52 molecule
- Antigen Symbol:CD52
- Clonality:Recombinant
- Clone:Campath-1H
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Cynomolgus monkey,Rhesus monkey,Human
- Western Blot:Yes
- Size:0.2 mg
- Cross Adsorption:No
- Form:Liquid
- Gene ID:1043
- Antigen Synonyms:CDW52|Cambridge pathology 1 antigen|Epididymal secretory protein E5|Human epididymis-specific protein 5|YTH66.9HL|Alemtuzumab
- Modification:Unmodified
- Storage Buffer:PBS with 0.02% Proclin 300
- Storage Temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Human lymphocytes.
- Tested Applications:Cytotoxicity assay, in vivo
- Cat. No.:76629-250
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: CD52 molecule
Clonality: Recombinant
Clone: Campath-1H
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG kappa
Reactivity: Human;Rhesus monkey;Cynomolgus monkey