Anti-NIP Recombinant Antibody [Clone: B1-8]
76629-240
: Prosci
- Antibody Type:Primary
- Clonality:Recombinant
- Clone:B1-8
- Conjugation:Unconjugated
- Isotype:IgG2b lambda
- Size:0.2 mg
- Cross Adsorption:No
- Form:Liquid
- Antigen Synonyms:NIP
- Modification:Unmodified
- Storage Buffer:PBS with 0.02% Proclin 300
- Storage Temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping Temperature:Shipped on blue ice
- Immunogen:hapten-protein conjugated NP12-chicken gammaglobulin (CG).
- Tested Applications:Negative control
- Cat. No.:76629-240
- Packaging:Vial
Type: Primary
Antigen: NIP
Clonality: Recombinant
Clone: B1-8
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG2b lambda
Reactivity: