Anti-CXCR2 Recombinant Antibody [Clone: HC2]
Catalog # 76630-190
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:C-X-C motif chemokine receptor 2
- Antigen Symbol:CXCR2
- Clonality:Recombinant
- Clone:HC2
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:0.2 mg
- Cross Adsorption:No
- Form:Liquid
- Gene ID:3579
- Antigen Synonyms:IL-8 receptor type 2|IL-8R B|CXC-R2|CD182|GRO/MGSA receptor|High affinity interleukin-8 receptor B|C-X-C chemokine receptor type 2|CXCR-2
- Modification:Unmodified
- Storage Buffer:PBS with 0.02% Proclin 300
- Storage Temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping Temperature:Shipped on blue ice
- Immunogen:This antibody was raised by immunizing mice with human IL-8 (interleukin-8) receptor, type B.
- Cat. No.:76630-190
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: C-X-C motif chemokine receptor 2
Clonality: Recombinant
Clone: HC2
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG1 kappa
Reactivity: Human