Anti-ADRB2 Recombinant Antibody [Clone: 1B1.3a]
Catalog # 76630-408
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:adrenoceptor beta 2
- Antigen Symbol:ADRB2
- Clonality:Recombinant
- Clone:1B1.3a
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Isotype:IgG kappa
- Reactivity:Mouse
- Size:0.2 mg
- Cross Adsorption:No
- Form:Liquid
- Gene ID:16154
- Antigen Synonyms:Interleukin-10 receptor subunit|CD210|Interleukin-10 receptor subunit alpha|CDw210a|IBI-3A|Il10ra Il10r|IL-10R subunit alpha|IL-10 receptor subunit alpha|1B1-3A|IL-10R subunit 1|Interleukin-10 receptor subunit 1|IL-10RA|IL-10R1
- Modification:Unmodified
- Storage Buffer:PBS with 0.02% Proclin 300
- Storage Temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping Temperature:Shipped on blue ice
- Immunogen:This antibody was raised by immunising rats with the purified recombinant extracellular region of mouse Interleukin 10 Receptor.
- Tested Applications:Blocking
- Cat. No.:76630-408
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: adrenoceptor beta 2
Clonality: Recombinant
Clone: 1B1.3a
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG kappa
Reactivity: Mouse