Anti-MHC class 1 Recombinant Antibody [Clone: R1-21.2]
76629-968
: Prosci
- Antibody Type:Primary
- Clonality:Recombinant
- Clone:R1-21.2
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b kappa
- Reactivity:Mouse
- Size:0.2 mg
- Cross Adsorption:No
- Form:Liquid
- Antigen Synonyms:H-2|MHC-I|R1.21.2|MHC Class I
- Modification:Unmodified
- Storage Buffer:PBS with 0.02% Proclin 300
- Storage Temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping Temperature:Shipped on blue ice
- Immunogen:This antibody was raised by immunising Lewis 1F rats with mouse CH-1 lymphoma cells. Spleen cells from immunised rats were fused with X-63Ag8-653 myeloma cells to generate hybridomas.
- Tested Applications:radioimmunoassay
- Cat. No.:76629-968
- Packaging:Vial
Type: Primary
Antigen: MHC class 1
Clonality: Recombinant
Clone: R1-21.2
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG2b kappa
Reactivity: Mouse