Anti-PD-1 Recombinant Antibody [Clone: RMP1-14]
Catalog # 76629-282
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Clonality:Recombinant
- Clone:RMP1-14
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- ImmunoChemistry:Yes
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Size:0.2 mg
- Cross Adsorption:No
- Form:Liquid
- Antigen Synonyms:RMP1/14|Programmed Death 1|RMPI-14|Programmed Death Receptor 1|CD279|PDCD1|PD1
- Modification:Unmodified
- Storage Buffer:PBS with 0.02% Proclin 300
- Storage Temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping Temperature:Shipped on blue ice
- Immunogen:The original rat hybridoma was generated by immunizing Sprague Dawley rats with mouse PD-1-transfectked BHK cells. A P3U1 myeloma was used as the fusion partner.
- Tested Applications:Blocking
- Cat. No.:76629-282
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: PD-1
Clonality: Recombinant
Clone: RMP1-14
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG2a kappa
Reactivity: Mouse